Probes and Seekers
Supplier: DR INSTRUMENTS INC. SE
35ANG 35H
470315-196BX 293.8 CAD
470315-196 470315-198
Probes and Seekers
Dissection Sets
Easily separate structures.
  • Hexagonal handle
Used to track vessels, nerves, and muscle fibers during dissection. Probes and seekers are a core dissection instrument. The blunt end causes minimal damage to nearby structures, while the pointed end can be used to tease apart tissue for access.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.