Iris Scissors
Supplier: DR INSTRUMENTS INC. SE
10 DR9TC
470315-164BX 203.8 CAD
470315-164 470315-234
Iris Scissors
Scissors
For fine dissections.
  • Used for very fine dissections, and working in small areas
The blades are very narrow allowing for a very clean cut, and a short blade length gives greater control. These iris scissors have excellent rust resistance. The scissors marked TC, are tungsten carbon tipped for greater edge retention and strength.
Ordering information: Available in curved or straight configuration.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.