MICROSTAR digital Series Electronic Overhead Stirrers, IKA
Supplier: IKA Works
0025004715 0025004883 0025004884
76329-844EA 2941.11 CAD
76329-844 76329-850 76329-942
MICROSTAR digital Series Electronic Overhead Stirrers, IKA
Stirrers Overhead Stirrers
The space-saving high performers MICROSTAR series features new and easy-to-use stirrers convince with perfect basic functionalities, as well as with a compact design.
  • Ease of operation takes place using a stable rotary knob
  • Display clearly shows the rotational speed
  • Regular software updates can be undertaken rapidly and simply via the USB interface
Compact design - The reduced design and the focus on the most essential aspects make this MICROSTAR stirrers reliable laboratory companions.
Constant torque - These stirrers guarantee a constant torque over the entire rpm range.