Rotational Viscometers, ROTAVISC, IKA
Supplier: IKA Works
113828 113829
76329-888EA 7198.83 CAD
76329-888 76329-890 76329-892 76329-894 76499-020 76499-022
Rotational Viscometers, ROTAVISC, IKA
Viscometers Rotational Viscometers
The ROTAVISC series determines the viscosity of liquids in all areas of application ranging from the laboratory to quality control. The four devices measure in different viscosity ranges.
  • Easy to operate
  • 4.3″ TFT display allows for an intuitive menu guidance
  • Digital spirit level supports the correct set-up of the start-up condition
  • Simple ramp function simplifies repetitive tasks
  • Stepless speed adjustment
Regardless of a simple or demanding viscosity measurement - the ROTAVISC delivers rapid and accurate results.
The level of measurement accuracy of the ROTAVISC rotational viscometer for both Newtonian and non-Newtonian fluids is +/- 1 % of the measuring range. The reproducibility is +/- 0.2 %.
Certifications: IP 40 protection class according to DIN EN 60529.
Delivery information: The scope of delivery includes a standard spindle set (SP1-SP4 or SP7-SP12), a protective bracket, temperature sensor and ROTASTAND stand.