VWR® T Balances, Legal-for-Trade (NTEP)
Supplier: VWR
VWR®
VWR-1003TC.N VWR-4502TC.N VWR-310TC.N VWR-6002TC.N VWR-160TC.N VWR-363TC.N VWR-205TC.N VWR-8100TC.N VWR-105TC.N VWR-2102TC.N VWR-4500TC.N VWR-6100TC.N VWR-220TC.N
76329-392EA 6716.94 CAD
76329-392 76329-396 76329-394 76458-450 76458-454 76458-444 76458-452 76458-442 76458-448 76458-456 76458-446 76329-400 76329-398
VWR® T Balances, Legal-for-Trade (NTEP)
Balances and Scales
The VWR® T-Series balances with NTEP certification feature state-of-the-art technology to provide the highest level of accuracy.
  • Class I and II options
  • Advanced features for weighing and documentation
  • Internal calibration
  • 2 x RS-232, wireless, ethernet, and USB (A & B) communication interfaces
  • Touchless operation via IR proximity sensors
  • 5-year warranty
These Class I (d=0.01 mg and 0.1 mg) and Class II (d=0.001 g and 0.01 g) balances offer the highest precision in legal-for-trade applications. The VWR® T-Series balances offer a variety of standard features, such as internal calibration, GLP procedures, adjustable ambient condition settings, 11 application modes, and multiple weighing units. The VWR® T-Series balances featuring an NTEP Certificate of Conformance to meet an organizations legal-for-trade requirements.
Certifications: CE Marked
Accessories information: Optional accessories such as printer (10799-132) and granite platform (10799-162) are available to order separately.
Ordering information: The 0.01 mg and 0.1 mg readability models feature a 3-door draft shield (which can not be removed). The 0.001 g readability models feature a removable glass draft shield. All units are supplied with switching AC adapter, weighing pan, weighing pan supports, user manual, and quick start guide. Accessories not included.
Caution: Fragile. Handle with care.