Manganese(II) sulfate monohydrate 98,0-102,0 Complexometric, Spray Dried Powder, EMPROVE® ESSENTIAL FCC, Ph. Eur., USP, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

Synonyms: Manganese sulphate monohydrate, Manganese sulfate monohydrate, Manganous sulfate monohydrate, Manganese(II) sulphate hydrate, Manganous sulphate monohydrate, Manganese(II) sulfate hydrate

1.05999.0500 1.05999.1000
CA1.05999.0500EA 355.66 CAD
CA1.05999.0500 CA1.05999.1000
Manganese(II) sulfate monohydrate 98,0-102,0 Complexometric, Spray Dried Powder, EMPROVE® ESSENTIAL FCC, Ph. Eur., USP, SAFC®
Manganese (II) sulphate monohydrate
Formula: MnSO₄·1H₂O
MW: 169.02 g/mol
Melting Pt: 700
Storage Temperature: Ambient
MDL Number: MFCD00149159
CAS Number: 10034-96-5
EINECS: 232-089-9
UN: 3077
ADR: 9 III
Merck Index: 13,05763

Order Now

Specification Test Results

Assay (Complexometric, MnSO₄·H₂O) 98.0 - 102.0%
Assay (Complexometric, Calc. on the Ignited Basis) 99.0 - 101.0%
Identity Passes Test
Appearance of Solution Passes Test
Chloride (Cl) ≤0.005%
Heavy metals (as Pb) ≤0.002%
As (Arsenic) ≤0.0003%
Ca (Calcium) ≤0.01%
Co (Cobalt)* ≤0.0005%
Fe (Iron) ≤0.001%
Ni (Nickel)* ≤0.0015%
Pb (Lead) ≤0.0004%
Se (Selenium) ≤0.003%
Zn (Zinc) ≤0.005%
Substances not Precipitated by Ammonium Sulfide (as SO₄) ≤0.5%
Residual solvents (Ph. Eur./USP/ICH) Excluded by Manufacturing Process
Loss on Ignition (500 °C) 10.5 - 12.0%
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Conforms to Ph. Eur., USP, FCC.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR