Sodium hydroxide 1 mol/L, EMPROVE® EXPERT
Supplier: EMD MILLIPORE # PCS CA EXCEP
|
Danger
|
1.37031.2500
1.37031.2500
1.37031.9025
1.37031.9200
EM1.37031.2500CS
1177.1
CAD
EM1.37031.2500
CA1.37031.2500
CA1.37031.9025
CA1.37031.9200
Sodium hydroxide 1 mol/L, EMPROVE® EXPERT
Sodium hydroxide
Formula:
NaOH Density: 1.04 g/cm³ (20 °C) |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1824 ADR: 8, |
Specification Test Results
Assay (Acidimetric, NaOH) | 0.99 - 1.01 mol/l |
Carbonate (as Na₂CO₃) | ≤1.0% |
Chloride (Cl) | ≤5 ppm |
Sulfate (SO₄) | ≤30 ppm |
Heavy Metals (as Pb) | ≤5.0 ppm |
Al (Aluminium) | ≤10 ppm |
Fe (Iron) | ≤3 ppm |
Residual Solvents (ICH Q3C) | Excluded by Production Process |
Endotoxins | ≤2.0 I.U./ml |
Total Aerobic Microbial Count (TAMC) | ≤100 CFU/g |
Total Combined Yeasts/Moulds Count (TYMC) | ≤10 CFU/g |
Filtered with 0.2 µm | |
This product is manufactured from sodium hydroxide pellets Ph. Eur., BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph. Eur. | |
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*). |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...