VWR® A Balances Legal-for-Trade (NTEP)
Supplier: VWR International
76329-402 76329-404 76329-406 76329-408 76329-410
76329-402EA 5448.16 CAD
76329-402 76329-404 76329-406 76329-408 76329-410
VWR® A Balances Legal-for-Trade (NTEP)
Balances and Scales
The A-series balances with NTEP certification feature state-of-the-art technology to provide the highest level of accuracy.
  • Class I and II options
  • Advanced features for weighing and documentation
  • Internal calibration
  • 2× RS232, USB (A and B), and Wi-Fi communication interfaces
  • Easy-to-read, adjustable, backlit display
The VWR® A-Series Balances with NTEP Certification offer the highest level or precision in legal-for-trade applications. These Class I and Class II certified balances feature an NTEP Certificate of Conformance indicating the devices meetMore Product Information