Specimen Containers, StarCLICK100™, Starplex Scientific
Supplier: Starplex
B1002-OCC B1002-YCC
CA76175-624CS 454.48 CAD
CA76175-624 CA76177-790
Specimen Containers, StarCLICK100™, Starplex Scientific
Sample Containers General Purpose Containers
When leakproof transport is essential, the solid, straight wall design of the container paired with the durable cap creates the optimal fit for preventing costly spills and biohazardous contamination.
  • Clear graduations on two sides in ounces and millilitres
  • Wide mouth design
  • Stackable
Correct cap closure is encouraged by listening for the double click confirmation during cap application.