Axygen® MultiRack Reloads, Corning
Supplier: Corning
MR-10XT-RL MR-200-RL MR-300-RL MR-1000XT-RL
76320-714CS 392.5 CAD
76320-714 76320-716 76320-718 76320-722
Axygen® MultiRack Reloads, Corning
Pipette Tips
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack reload system is designed to fit Axygen® MultiRack as well as racks from other suppliers without tedious realignment. This design not only saves labMore Product Information