Axygen® Linear Stands for Four or Six Pipettors, Corning
Supplier: Corning
AP-USD4-P AP-USD6
76324-848CS 145.69 CAD
76324-848 76324-852
Axygen® Linear Stands for Four or Six Pipettors, Corning
Pipette Stands
Universal linear stands for up to four or six pipettors.
Red model: For up to four single-channel pipettors or maximum two 8-channel or 12-channel pipettors.
Transparent model: For up to six single-channel pipettors or maximum three 8-channel or 12-channel pipettors.