FlashPure EcoFlex Cartridges, Büchi
Supplier: Buchi
140000048 140000049 140000050 140000051 140000052 140000053 140000054 140000055 140000056 140000057 140000058 140000059 140000060 140000061 140000062 140000063 140000064 140000065 140000066 140000067 140000068 140000069 140000070 140000071 140000072 140000073 140000074 140000098 140000102 140000021 140000022 140000023 140000024 140000025 140000026 140000027 140000028 140000029 140000030 140000031
76196-780BX 368.44 CAD
76196-780 76196-782 76196-784 76196-786 76196-788 76196-790 76196-792 76196-794 76196-796 76196-798 76196-800 76196-802 76196-804 76196-806 76196-808 76196-810 76196-812 76196-814 76196-816 76196-818 76196-820 76196-822 76196-824 76196-826 76196-828 76196-830 76196-832 76196-834 76196-836 76196-838 76196-840 76196-842 76196-844 76196-846 76196-848 76196-850 76196-852 76196-854 76196-856 76196-858 76196-860 76196-862 76196-864 76196-866 76196-868 76196-870 76196-872 76196-874 76196-876 76196-878 76196-880 76196-882 76196-884 76196-886 76196-888 76196-890 76196-892 76197-010 76197-012 76197-014 76197-016 76197-018 76197-020 76197-022 76197-024 76197-026 76197-028 76197-030 76197-032 76197-034 76197-036 76197-038 76197-040 76197-042 76197-044 76197-046 76197-048 76197-050 76197-052 76197-054 76197-056 76197-058 76197-060 76197-062
FlashPure EcoFlex Cartridges, Büchi
Chromatography Columns
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries. This enables the user to choose the flash cartridge which best suits his purification needs.
  • Different sizes for scalability from milligrams to grams
  • Broad range of different normal and reversed phases
  • Small and spherical particles for fast separations and high loadings
Compatibility with the Pure Navigator software (flash method optimization by interpreting TLC or HPLC scouting runs). RFID tag for an automatic transfer of important parameters for a run on a Pure chromatography system.
Silica phase are high and medium polar compounds, most popular polar phase, mainly used for flash applications. C18 WP (wide pores) has a ideal application in peptides.
C18 phase are low polar and non-polar compounds, most popular non-polar phase. Amino phase are high and medium polar molecules example carbohydrates and amines. Alumina Wide variety of molecules example acidic, basic or acid sensitive compounds.
Diol, C18 and Amino phase has spherical particle shape. Silica and alumina has irregular shape.
Loading capacity: 0.1 to 10% for Silica, Silica LOK, Silica Solid Loader; 0.1 to 4% for Alumina Neutra, Alumina Basic, Alumina Acidic; 0.1 to 2.5% for C18; 0.1 to 5% for Amino, Diol.