Rectangular and Square Cover Slips, Bellco Glass
Supplier: Bellco Glass
1916-09009A 1916-09022 1916-09035A 1916-09050A 1916-10522A 1916-10535A 1916-10550A 1916-12222A 1916-13054A 1916-24050A 1916-25075
76305-488CS 88.32 CAD
76305-488 76305-490 76305-492 76305-494 76305-496 76305-498 76305-500 76305-502 76305-504 76305-506 76305-508
Rectangular and Square Cover Slips, Bellco Glass
Cover Glasses
These cover slips provides good quality, reliability, and better performance for all laboratory microscopy needs
  • #1 thickness (0.13 to 0.16 mm)
  • Packed 100 per case
Clear white borosilicate glass.
Order Now
SPECIFICATIONS
Environmentally Preferable