Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V, British (UK), Labconco
Supplier: Labconco
302411151 302580151 302380051 302380151 302381051 302381151 302410051 302410151 302411051 302480051 302481051 302481151 302510051 302510151 302511051 302580051 302581051 302581151 302610051 302610151 302611051 302611151 302680051 302680151 302681051 302681151 302480151 302511151
76319-136EA 23052.2 CAD
76319-136 76319-150 76319-164 76319-184 76319-194 76319-208 76319-216 76319-230 76319-244 76319-258 76319-266 76319-280 76319-294 76319-310 76319-326 76319-340 76319-354 76319-368 76319-382 76319-404 76319-420 76319-434 76319-448 76319-462 76319-476 76319-490 76319-504 76319-518 76319-526 76319-540 76319-562 76319-568
Purifier® Logic®+ Class II A2 Biosafety Cabinets, 230 V, British (UK), Labconco
Fume Hoods and Enclosures Biological Safety Cabinets
Purifier Logic+ Type A2 biosafety cabinets combine ergonomics, comfort and safety.
  • MyLogic OS displays cabinet status and alarms with clear, concise images and messaging to build user confidence
  • Constant Airflow Profile™ (CAP) Technology automatically maintains a precise volume of air as the filters load to protect a user and their samples
  • Ergonomic Inclination™ Technology Suite includes 10° angled sash, patented curved air grille with built-in armrest, fully accessible controls and eye-level mounted color display
  • Quiet, efficient ECM blower and energy saving LED lights reduces cabinet’s carbon foot print
  • Provides personnel, product and environmental protection and is suitable for work in all biosafety levels
The Purifier Logic+ Class II A2 Biosafety Cabinet excels in user comfort and functionality. Industry leading advances in ergonomic design have shaped the Inclination™ Technology Suite that includes the following: 10° angled sash, LED lighting, patented curved air grille with built in armrest, fully accessible controls and line-of-sight mounted color display with MyLogic™ operating system.
MyLogic OS provides intuitive control of the Logic+ with clear messages and vibrant imagery, status updates and alarms with diagnostic troubleshooting. Using ECM blower technology, the Logic+ uses the Constant Airflow Profile™ (CAP) to maintain a precise volume of air as the filters load. In addition, the cabinet will continuously display the HEPA filter life remaining and cabinet status. These features combine to provide unparalleled safety.
Type A2 biosafety cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels. These biosafety cabinets are designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through an efficient Ventus™ Canopy Connection (optional). In addition, some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included.
Certifications: ETL listed. NSF listed. ADA-compliant (height of controls and receptacles). CE Conformity marking (230 V models). ISO 5 conditions per ISO 14644-1 and 2.
Accessories information: Accessories not included unless stated.
Ordering information: Base stand required if model with stand is not selected.