Tributylammonium pyrophosphate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Tributylammoniumpyrophosphate, Bis(tri-n-butylammonium) pyrophosphate

T2047-250MG T2047-5GM
76026-486EA 184.39 CAD
76026-486 76026-488 76026-490
Tributylammonium pyrophosphate
Tributylammonium pyrophosphate
Formula: C₂₄H₅₈N₂O₇P₂
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD00136019
CAS Number: 5975-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR