Thiourea ACS

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Sulfourea, Thiocarbamide, Sulphourea

T1040-18 T1040-10
CA11020-404EA 4665 CAD
CA11020-404 CA11024-432
Thiourea ACS
Thiourea

Thiourea, Crystal, Reagent, ACS is an organosulfur compound that is structurally similar to urea. Used in organic synthesis to reduce peroxides to the corresponding diols, it can also be applied in the reductive workup of ozonolysis to then give carbonyl compounds. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: NH₂CSNH₂
MW: 76.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008067
CAS Number: 62-56-6
UN: 2811
ADR: 6.1 III
Merck Index: 13,09443
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fff6222c54fe16ea4d36edcbf320745cb1e18d6&vid=e9ae5dc49e92d68f893244a5cd4e0245f289ed09&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR