Sodium chlorate, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

S1237-12KG S1237-500GM S1237-2.5KG
76264-480EA 1266.29 CAD
76264-480 76264-516 76264-518
Sodium chlorate, Technical Grade
Sodium chlorate

Sodium Chlorate, Technical, considered hygroscopic, is used in the applications of bleaching paper.


Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Formula: ClNaO₃
MW: 106.44 g/mol
Boiling Pt: 300
Melting Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00003479
CAS Number: 7775-09-9
UN: 1495
ADR: 5.1 II
Merck Index: 13,08670

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR