Orthophosphoric acid 25% (v/v) in aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Phosphoric acid

P-370-52
CA11024-394EA 376.17 CAD
CA11024-394
Orthophosphoric acid 25% (v/v) in aqueous solution
Orthophosphoric acid

Phosphoric Acid, 25 Percent (v/v) Aqueous - Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in LabChem’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability

Formula: H₃PO₄
MW: 98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR