Mercury(II) nitrate solution 0.01 N in aqueous solution

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Mercury dinitrate, Mercuric nitrate

M-174-52
CA11020-402EA 366.01 CAD
CA11020-402
Mercury(II) nitrate solution 0.01 N in aqueous solution
Mercury (II) nitrate

Mercuric Nitrate, 0.01 N Aqueous Solution, also known as mercury(II) nitrate solution, acts as an oxidizer in analytical chemistry applications. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability

Formula: HgN₂O₆
MW: 324.6
Melting Pt: -38.9
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00011038
CAS Number: 10045-94-0
Merck Index: 12,05933

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR