Cesium nitrate, Ultrapure
Supplier: SPECTRUM CHEMICAL MFG CORP CA
Synonyms:
Nitric acid cesium salt
C1489-05
CA11024-302EA
551.1
CAD
CA11024-302
Cesium nitrate, Ultrapure
Cesium nitrate
Cesium Nitrate, Ultrapure is used as a colorant, as an oxidizer, and in pyrotechnic compositions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
CsNO₃ MW: 194.91 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010963 CAS Number: 7789-18-6 UN: 1451 ADR: 5.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...