Ammonium sulfide solution ACS
Supplier: SPECTRUM CHEMICAL MFG CORP CA
Ammonium Sulfide Solution, Reagent, ACS is an unstable salt with the formula (NH4)2S. Common uses of AMMONIUM SULFIDE are industrial applications including photo developing and textile manufacturing.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
H₈N₂S Density: 0.997 Storage Temperature: Ambient |
MDL Number:
MFCD00010892 CAS Number: 12135-76-1 UN: 2683 ADR: 86.13,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...