Sodium deoxycholate ≥96% (by TLC), Calbiochem®, Millipore®

Supplier: MilliporeSigma
Warning

Synonyms: Deoxycholic acid sodium salt, 3α,12α-Dihydroxy-5β-cholan-24-oic acid sodium salt, Sodium 3α,12α-dihydroxy-5β-cholanoate

264101-1KG 264101-25GM
CA80058-004EA 4218.83 CAD
CA80058-004 CA80058-006
Sodium deoxycholate ≥96% (by TLC), Calbiochem®, Millipore®
Sodium deoxycholate

An ionic detergent for the solubilization of membrane-bound proteins.

Formula: C₂₄H₃₉NaO₄
MW: 414.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064139
CAS Number: 302-95-4

Order Now

Specification Test Results

Purity ≥96% (TLC)
Heavy metals ≤20.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR