Mini Vortex Mixers with Touch Function

Supplier: Heathrow Scientific
120598 120567
470316-814CS 2736.55 CAD
470316-814 470316-828
Mini Vortex Mixers with Touch Function
Shakers Vortex Mixers
A simple, touch start operation enables exceptional mixing performance with one handed operation.

  • Made of durable ABS and silicone materials to resist most commonly used chemicals
  • Rapid vortex action saves time and enables quick mixing of samples
  • A low activation force helps prevent user fatigue on repetitive procedures
  • Stable platform
  • Quiet operation and low vibration
  • Can be used with tubes ranging from 2 to 50 ml

Low profile design and small footprint (only 4.3" wide) save valuable space on a bench top and in fume hoods. Easy one handed touch start operation and shaking stops when the tube is lifted.

Certifications: CE, CB Scheme, and SGS (Canada and U.S.)

Ordering information: Five-year warranty.

Delivery information: Includes one mini vortexer, two silicon caps, one low voltage double insulated power adapter with four interchangeable plugs.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR