Magnetic Induction Stirrers
Supplier: Heathrow Scientific
120585 120584
470316-810CS 1562.6 CAD
470316-810 470316-812
Magnetic Induction Stirrers
Stirrers Magnetic Stirrers
This magnetic induction stirrer is motor-less, using advanced coil technology that produces the strongest magnetic coupling and has no moving parts making this unit 100% wear-free and maintenance free. The magnetic coil technology works on the inductive principle with alternative current (AC) as its driving force.
  • No heat is generated after hours of continuous use
  • Ideal for temperature sensitive samples and environments
  • Quiet operation and low vibration
  • The magnetic field driving the stir bar maintains continuous constant speeds for reproducible results
  • Speed remains constant,even when load changes
  • Rapid acceleration and braking, less than ten seconds
  • An easy to read digital display precisely controls time (30 seconds to 60 minutes) and speed (50 to 2000 rpm)
  • Durable ABS housing is corrosion resistant and easy to clean
This uses advanced coil technology resulting in the unit creating the strongest magnetic coupling, perfect for viscous solutions.
Once a program has started, the speed, cycle and time can be monitored. The stirring speed can be adjusted while a program is running.
Stirrer bars are coated with PTFE for chemical resistance. Stirrer modes: Single clockwise, single counter clockwise, auto reverse rotation.
Mixing Surface is 165 mm diameter, silicone mat is 160 mm in diameter.
Certifications: CE, SGS, RoHS2, WEEE.
Ordering information: Five-year warranty.
Delivery information: Includes two stir bars (35×12 mm), one silicon anti-skid plate cover, one low voltage, double insulated power adapter with four interchangeable plugs.