ChemRxnHub™ Systems, Jacketed, 500 ml, Jacketed Lab Reactor, Chemglass
Supplier: Chemglass
CG-1949-X-500 CG-1949-X-500M CG-1949-X-505M CG-1949-X-505
30622-582EA 26114.65 CAD
30622-582 30622-584 10170-678 10171-106
ChemRxnHub™ Systems, Jacketed, 500 ml, Jacketed Lab Reactor, Chemglass
Laboratory Reactor Systems
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.