ChemRxnHub™ Systems, Jacketed, 1000 ml, Jacketed Lab Reactor, Chemglass
Supplier: Chemglass
CG-1949-X-1L CG-1949-X-1LM
30622-570EA 26347.54 CAD
30622-570 30622-572
ChemRxnHub™ Systems, Jacketed, 1000 ml, Jacketed Lab Reactor, Chemglass
Laboratory Reactor Systems
ChemRxnHub™ process systems are complete with benchtop stand, overhead stirrer and glassware.
Available with the optional manifold connection and heat transfer fluid draining system. The unique design of the ChemRxnHub™ support stand and clamping system allows for quick and easy installation/removal of glassware and fits into most benchtop hoods.