Formamidine hydroiodide, low water content ≥98.0%

Supplier: TCI America

Synonyms: FAI, Formamidinium iodide

F0974-1G F0974-5G F0974-25G
TCF0974-1GEA 105.86 CAD
TCF0974-1G TCF0974-5G TCF0974-25G
Formamidine hydroiodide, low water content ≥98.0%
Formamidine hydroiodide
Formula: CH₄N₂·HI
MW: 171.97 g/mol
MDL Number: MFCD28369273
CAS Number: 879643-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a67dd44ecd31d29488517c33c553ec51e098536&vid=3a67dd44ecd31d29488517c33c553ec51e098536&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity(with Total Nitrogen) min. 98.0 %
Purity(Argentmetric Titration) min. 98.0 %
Water max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR