Replacement Tips, Excelta Corp®
Supplier: Excelta
159D-RTX 159-RTX 159A-RTX 159B-RTX 159C-RTX 159E-RTX 159F-RTX 159G-RTX
89020-044EA 25.53 CAD
89020-044 94002-670 94002-672 94002-674 94002-676 94002-678 94002-680 75880-638 76471-538 76471-542
Replacement Tips, Excelta Corp®
Forceps
High-quality forcep tips.
  • High rigidity, excellent tensile and flexural strength.
These soft replacement tips are made of a Carbofib™ material which is a polymer based material with 30% carbon fibers added. The working temperature is 249 °F. Carbofib™ has very high rigidity, excellent tensile and flexural strength, and fatigue and creep resistance. The tips are conductive (102 ohms/sq). Each includes 4 screws and 2 tips.
Order Now
SPECIFICATIONS
Environmentally Preferable