Cutters, Transverse End, Excelta
Supplier: Excelta
7274E 7173E 7273E
63042-812EA 115.56 CAD
63042-812 94024-370 75880-694 75880-702
Cutters, Transverse End, Excelta
Cutters
Thes cutters are ideal for horizontal cutting on a vertical plane in tight places.
  • Transverse cutting blades for difficult areas
  • 'Set-Screw' stop prevents over-travel
  • Induction hardened blades for longevity
  • Ergonomic handles and dual leaf springs
These Lazer Line® miniature tapered transverse optimum flush cutters with a cutting capacity of 0.001 to 0.020" soft wire is made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. This 6" tool has a 40°More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable