UVP Pen-Ray® Field Packs, Analytik Jena
Supplier: Analytik Jena US
95-0256-01 95-0256-02
21474-562EA 1128.17 CAD
21474-562 21474-564
UVP Pen-Ray® Field Packs, Analytik Jena
Lights UV Lights, Lamps and Bulbs
Pen-Ray® Field Packs combine a Pen-Ray® UV light source and a DC power supply (with 9 V battery) in a compact, portable package.
  • Field Packs are lightweight (0.3 lbs.) and small enough to fit in a pocket or be carried by the belt clip attached to the power supply
  • Field Packs are available with a choice of options for customization or in three standard configurations
  • Field Pack FP-L/S provides two interchangeable light sources (365 and 254 nm); a lamp connector is located on the outside of the power supply for this configuration only
  • Other configurations
More Product Information