Soothers™ Moisturizing Disposable Earplugs, Moldex®

Supplier: Moldex
Soothers™
6680
76326-010BX 117.43 CAD
76326-010
Soothers™ Moisturizing Disposable Earplugs, Moldex®
Earplugs Disposable Earplugs
Moldex® Soothers™ foam earplugs are made of extra-soft, extra light foam. The design is so comfortable, users forget they’re wearing earplugs.

  • Moisturizes for all day comfort
  • Soft foam and tapered shape for added comfort
  • Highest independently tested NRR 33 rating
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Shaped to fit even very small ear canals
  • 100% PVC-free for a greener alternative

Soothers™ earplugs offer a high NRR of 33dB, making them suitable for most noisy environments. The tapered shape fits easily into the ear canal, providing a gentle and snug seal without uncomfortable pressure. Plus, the outer surface of the earplug is smooth, and does not irritate the ear canal.

Ordering information: Order requires a 6 day lead time.

Packaging: Earplugs are packaged 200 pairs per box.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR