Notrax® 926 Smooth Stat™ Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
926S0035BL 926S0023BL
76310-738EA 587 CAD
76310-738 76310-910
Notrax® 926 Smooth Stat™ Floor Mattings, Justrite®
Floor Mats Tacky Mats
Smooth Stat™ is a dissipative/anti-static mat designed to absorb static electricity.
  • Electrically conductive PVC smooth top surface
  • Dense closed cell PVC foam base
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all 4 sides
  • Certified by the NFSI (National Floor Safety Institute)
  • Grounding cord included, required
Mats drain static electricity from the worker, keeping sensitive circuitry from being damaged. The durable vinyl smooth top surface provides comfort and ergonomic support andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable