Notrax® 539 WebTrax® Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
539S0035GY 539S0035BU 539R0340GY 539R0340BU 539R0440BU 539R0440GY
76310-740EA 573.1 CAD
76310-740 76310-742 76310-828 76310-830 76310-840 76310-842
Notrax® 539 WebTrax® Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
WebTrax® is a multi-functional vinyl floor mat that can be used in a variety of wet applications.
  • Industrial grade extruded PVC
  • Open construction of extruded vinyl has a wave design that creates a slip-resistant walking surface
  • Oil-, grease-, and UV-resistant
  • Performs well in different climates and adapts to any surface
  • Free of: DOP, DMF, silicone, heavy metal, ozone depleting substances
  • Factory compressed borders
The wave pattern design creates a slip-resistant walking surface that increases traction on wet and slippery floors. Antibacterial treated PVC material prevents the growth of fungi or bacteria while the open structure design allows for easy draining of liquids. The industrial grade PVC compound also provides greater resistance to caustic chemicals, oils, and greases.
Material PVC