Notrax® T17 Superfoam™ Floor Mattings, Justrite®
Supplier: SUPERIOR MANUFACTURING GROUP INC
NoTrax®
T17S0033BL T17P0036BL T17S0038BL T17P0034BL T17P0035BL T17S0032BL T17C0036BL T17C0048BL T17P0032BL T17S0036BL T17P0033BL T17S0035BL T17P0038BL T17S0034BL
76310-648EA 181 CAD
76310-648 76310-968 76310-948 76310-662 76310-762 76310-696 76310-784 76310-774 76310-730 76310-920 76310-734 76310-548 76310-550 76310-694
Notrax® T17 Superfoam™ Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Superfoam™ is an extremely lightweight and durable anti-fatigue foam mat designed specifically for ease of handling and ergonomic comfort.
  • Closed cell nitrile foam, does not absorb liquids
  • Durable and abrasion resistant
  • Solid for dry areas
  • Perforated drainage holes to remove liquid and debris for worker platform
  • Non-allergenic, silicone-free compound
  • 45° beveled edges on all four sides
Made from a closed cell PVC Nitrile foam blend, Superfoam™ does not absorb liquids and is highly resistant to greases, oils, animal and vegetable fats. This allows it to be used in areas where occasional overspray and dripping occur, such as dishwashing stations and bar areas. Because Superfoam™ is lightweight, it is the perfect mat for use in areas where mats are regularly moved for cleaning like cooking lines, service counters, and food prep stations.
Material: Sponge/Rubber