Notrax® 419 Diamond Sof-Tred™ with Dyna-Shield® Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
419S0035BY 419R0036BL 419R0048BL 419R0024BY 419R0036BY
76310-626EA 159.63 CAD
76310-626 76310-808 76310-816 76310-984 76311-000
Notrax® 419 Diamond Sof-Tred™ with Dyna-Shield® Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Engineered for ultimate comfort and safety.
  • Non-directional diamond plate textured embossed mat
  • PVC foam provides added cushioning comfort
  • Exclusive Dyna-Shield® top surface manufactured directly into the mat for long lasting wear
  • Superior tear resistance and tensile strength
  • Beveled edges on all 4 sides
Diamond Sof-Tred™ with Dyna-Shield® anti-fatigue matting is designed to provide traction with a non-directional diamond plate embossed top surface. This allows for sure footing and is easy to sweep clean. The exclusive Dyna-Shield® PVC sponge isMore Product Information