Notrax® 951 Knee RX™ Heavy-Duty Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
951S1222BL 951S4122BL
76310-590EA 114.6 CAD
76310-590 76310-684
Notrax® 951 Knee RX™ Heavy-Duty Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Knee RX™ Heavy-Duty mats are designed for extreme or extended kneeling on surfaces such as gravel, asphalt, concrete, tile, and wood flooring.
  • Designed for comfort and durability in extreme kneeling applications
  • For use on gravel, asphalt, concrete, dirty/oily surfaces, tile and wood flooring, metal grating, or grass
  • Durable laminated heavy-duty rubber bottom surface for additional comfort and long-lasting wear
  • Nitrile/PVC foam blend for the highest degree of resistance to oils and greases
  • Self-extinguishing, non-conductive, silicone-free material
  • Built-in handle easy to carry or hang up
  • 1.83 sq. ft. coverage area
Mats provide ergonomic support by reducing trauma, stress, and strain on the knees and lower back. Perfect for tool and maintenance carts, or for individual workstations, Knee RX™ is made from a nitrile/PVC foam blend offering resistance to oils and greases. It is a full 1” thick for the ultimate in comfort. A durable, heavy-duty rubber bottom surface ensures long-lasting wear in the roughest environments. Knee RX™ HD provides a comfortable work surface for those who are required to occasionally kneel while working and eliminates the hassle and discomfort of wearing knee pads.
Material: Laminate/Rubber