Notrax® 782 Bubble Trax® Ultra™ Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
782S0023BL 782S0035BL 782S0023GY 782S0023BY 782S0035GY 782S0035BY
76310-664EA 198.05 CAD
76310-664 76310-732 76310-856 76310-858 76310-868 76310-870
Notrax® 782 Bubble Trax® Ultra™ Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
Bubble Trax® Ultra™ is a unique, high-performance, anti-fatigue floor mat with a bubble top surface design providing extreme ergonomic comfort and durability.
  • Heavy-duty vinyl bubble top surface
  • Dense, closed cell PVC foam base provides added cushioning comfort
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all four sides
  • Certified by the NFSI (National Floor Safety Institute)
Bubble Trax® combines a heavy-duty vinyl top surface with a dense sponge backing utilizing UniFusion™ technology. This virtually eliminates the possibility of de-lamination. All Notrax® laminate floor mats come standard with RedStop™, a uniquely engineered non-slip backing technology that eliminates slipping and sliding on smooth surfaces such as tile, wood, marble, and treated concrete.