Notrax® 457 Skystep™ ESD Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
457S0023BL 457S0035BL 457S0034BL
76310-628EA 165.57 CAD
76310-628 76310-692 76310-958
Notrax® 457 Skystep™ ESD Floor Mattings, Justrite®
Floor Mats Traction and Anti-Fatigue Floor Mats
The Skystep™ ESD static dissipative mat is a stand-alone rubber mat designed for individual work stations. Ergonomically engineered, the SkyStep™ ESD utilizes the power of air.
  • Superb anti-fatigue properties due to the ergonomic bubble pattern
  • Rubber molded, air-pocket underside anti-fatigue mat
  • Made of heavy duty natural rubber compound
  • ESD Static Dissipative, measured resistance 105 to 107 Ω
  • Beveled edges on all four sides
  • Grounding cord available
The mat is designed to suction to the ground to prevent shifting while trapping air in its chambers.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable