Anticon® MicroQuilt Wipes, Contec®
Supplier: CONTEC, INC. MX
495758-735 495758-736 495758-737 495758-738
76325-986CS 1196.4 CAD
76325-986 76325-988 76325-990 76325-992
Anticon® MicroQuilt Wipes, Contec®
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Anticon® MicroQuilt combines the cleanliness and sorbent capacity of Quiltec® with the microfiber particle pick-up ability of Anticon® Gold Sorb. This hybrid creates the most sorbent, cleanest wipe Contec® offers.
  • Hybrid of polyester and microfiber for the most sorbent and clean wipe Contec® offers
  • Laminated two-ply wipe readily releases fluid to a surface
  • Unique microfiber polyester yarn offers high sorptive capacity and fast absorption rate
  • Sealed edges reduce particle and fiber generation
MicroQuilt is laser-cut with heatsealed edges and cleanroom laundered to reduce the number of releasable particles and fibers. The unique microfiber construction provides superior residue and particle removal when used dry or wet. The 2-ply, quilted layers absorb more liquid than other wipes but readily release fluids when applying to surfaces. Composed of microfiber polyester, the wipes are durable and compatible with a wide range of solutions and disinfectants. Anticon® MicroQuilt is ideal for any cleanroom application requiring high sorbent capacity and superior contaminant removal.