Wright's stain, powder, HARLECO® stain for hematology, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Eosin Methylene blue solution according to Wright
, Wright's eosin methylene blue
376-12
376-12
376-16
376-29
376-29
376-16
CA34172-114EA
4911.87
CAD
34172-114
CA34172-114
15204-110
34172-076
CA15204-110
CA34172-076
Wright's stain, powder, HARLECO® stain for hematology, Sigma-Aldrich®
Wright's stain
For differential blood staining.
- For In-Vitro Diagnostic Use.
- For Professional Use.
Certified by the Biological Stain Commission (BSC).
Storage Temperature:
Ambient |
MDL Number:
MFCD00082143 CAS Number: 68988-92-1 EINECS: 273-541-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...