Biologix dPette Digital Pipettes
Supplier: TWD Scientific
BX01-2441 BX01-2431 BX01-2421 BX01-2411
76319-884EA 695.83 CAD
76319-884 76319-886 76319-888 76319-890
Biologix dPette Digital Pipettes
Pipettes Micropipettes
These high-quality digital pipettes feature built in error control and follow a comprehensive range of liquid handling protocols.
  • Three aspiration/ dispensing speeds
  • LCD screen
  • Motor drive with built in error control
  • Lithium-ion rechargable battery
  • Autoclavable lower barrel
dPette digital pipette features three dispening speeds and single-channel adjustable volume. Various sizes available.
Accessories information: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
Ordering information: Charging cord, charging base and power box included.Quality Control Certification Included.