Tin(II) chloride 40% hydrochloric solution

Supplier: Ricca Chemical

Synonyms: Stannous chloride, Tin dichloride

R7995000-1C R7995000-120C R7995000-250C R7995000-500C
RCR7995000-1CEA 632 CAD
RCR7995000-1C RCR7995000120C RCR7995000250C RCR7995000500C
Tin(II) chloride 40% hydrochloric solution
Tin (II) chloride
Formula: SnCl₂
MW: 189.62 g/mol
Boiling Pt: 623
Melting Pt: 246
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
UN: 1789
ADR: 8 II
Merck Index: 13,08861

Order Now

Specification Test Results

Appearance Colorless to Light Yellow-Green Passed Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR