Stainless Steel Clamps, Ace Glass
Supplier: Ace Glass
7600-30 7600-40 7600-05 7600-10 7600-15 7600-20 7600-25
89059-637PK 84.79 CAD
89059-637 89059-641 89059-627 89059-629 89059-631 89059-633 89059-635
Stainless Steel Clamps, Ace Glass
Clamps Standard Taper and Spherical Joint Clamps
These corrosion-free stainless steel spring wire clamps will securely hold 10/30 - 45/50 glass joints with no screw needed.
Can be used to secure thermometers, vacuum systems, distillation set-ups, chromatographic columns, evaporators, or any glass apparatus that has to be secured with springs or clamps. Be sure to use the proper clamp for the designated joint size.