Media Bottles, Square, PET, TriForest
Supplier: TriForest
PTC0125S-D PTC1000S-C PTC0125S-A PTC0060S-C PTC0250S-C PTC0650S-C PTC0650S-A PTC0030S-D PTC0060S-D PTC0250S-A PTC0060S-A PTC0030S-A PTC0125S-C PTC0250S-D PTC0650S-D
76320-304CS 351.09 CAD
76320-304 76320-348 76320-328 76320-356 76320-344 76320-358 76320-346 76320-330 76320-352 76320-362 76320-340 76320-354 76320-332 76320-342 76320-350
Media Bottles, Square, PET, TriForest
Bottles Plastic Bottles
Bottles are ideal for the storage of culture media, reagents, serum, and biological buffers.
  • Excellent gas barrier properties; reduced permeability to CO₂/O₂
  • Sterility of 10⁻⁶ SAL
  • Molded graduations for approximate volume reference
  • Non-pyrogenic, nuclease-free, and non-cytotoxic
  • Bottle and cap material are Certified Animal Derived Component-Free
Square polyethylene terephthalate bottles are available with or without high density polyethylene caps in shrink wrapped trays. Lightweight and shatter resistant, bottles are recommended for storage of all compatible solutions, biologicals, liquid culture media, and aqueous buffers.
Bottles are available with standard cap (PTCXXXXS-C), tamper evident cap (PTCXXXXS-A), or without cap (PTCXXXXS-N) for use with automated fill lines.
Certifications: Compliance with USP Class VI guidelines.