Advanced Counterstain, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
S128A
S008A
CA89407-356EA
471.22
CAD
CA89407-356
CA89407-358
Advanced Counterstain, Hardy Diagnostics
Stains
Microbiology Stains
Use Advanced Counterstain to stain gram negatives a vivid and distinct red. The clear advantage is that the gram negative organisms will appear brighter with better definition at the edges.