Sampling Bags, SteriBag Blue, Bürkle
Supplier: BURKLE INC
5344-7008 5344-7012
76319-738PK 911.38 CAD
76319-738 76319-740
Sampling Bags, SteriBag Blue, Bürkle
Bags Sample Bags
The sterile sampling bag SteriBag Blue is especially well suited for use in the food, animal feed and pharmaceutical industries. To prevent the danger of foreign matter in production, the sample bags and the tear-off closure are colored blue. This shade of blue does not occur in a natural form in foodstuffs. Because of this it is highly visible and can be reliably discovered in a visual check if a bag or parts of it are ever mixed into production.
  • PE, blue
  • Liquid-tight
  • With labeling field
  • Sterile, RNase, DNase and pyrogen-free
The disposable sampleMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable