Sampling Bags, SteriBag Cleanroom, Bürkle
Supplier: BURKLE INC
5344-8010 5344-8012
76319-742PK 1390.42 CAD
76319-742 76319-744
Sampling Bags, SteriBag Cleanroom, Bürkle
Bags Sample Bags
The sampling bag SteriBag Cleanroom with double packaging can be placed directly in the clean room. 25 sample bags are placed in double secondary packaging. They can be removed one after the other and are therefore ideally suited for introducing into the clean room. Then the sterile bags can be used directly in the clean room.
  • Sterile, with double packaging
  • RNase, DNase and pyrogen-free
  • PE, transparent
  • Liquid-tight
  • With labelling field
SteriBag Cleanroom is sterile inside and outside. This guarantees contamination-free storage and transport of solid, semi-solid and liquid media. The sample bag is perfect for use in chemical and pharmaceutical applications. The disposable sample bag has a labelling field, saves space and is also easy to transport. The large bag opening makes it easy to pour in the sample.