Sulfuric acid 95.0-98.0% (by alkalimetry), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, SAFC®

Supplier: MilliporeSigma
Danger

1.00713.2500 1.00713.9025 1.00713.2500
CA1.00713.2500CS 1548.87 CAD
CA1.00713.2500 CA1.00713.9025 EM1.00713.2500
Sulfuric acid 95.0-98.0% (by alkalimetry), EMPROVE® ESSENTIAL NF, Ph. Eur., BP, JPE, SAFC®
Sulfuric acid

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Order Now

Specification Test Results

Assay (Alkalimetric) 95.0 - 98.0%
Identity Passes Test
Appearance of Solution Passes Test
Chloride (Cl) ≤0.0002%
Nitrate (NO₃) ≤0.00010%
Nitrate (NO₃) (Ph. Eur.) Passes Test
Heavy Metals (as Pb) ≤0.0005%
As (Arsenic) ≤0.000010%
Fe (Iron) ≤0.0002%
NH₄ (Ammonium) ≤0.0005%
Pb (Lead) ≤0.0005%
V (Vanadium) ≤0.0002
Residual Solvents (ICH Q3C) Excluded by Production Process
Substances Reducing Potassium Permanganate (as SO₂) ≤0.0010%
Residue on Ignition ≤0.0050%
Corresponds to Ph. Eur., BP, JPE, NF


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR