BD Vacutainer® Luer-Lok™ Access Device, BD
Supplier: BD Biosciences
Luer-Lok®
36490200
VT36490200CS 464.86 CAD
VT36490200
BD Vacutainer® Luer-Lok™ Access Device, BD
Blood Collection Tube Holders
The BD Vacutainer® Luer-Lok™ access device provides sterile, secure and safe specimen sampling for fast, accurate and consistent results that can lead to improved patient outcomes.
  • Sterile
  • Latex-free
  • Red colored tip for differentiation
It offers the security of a threaded, Luer locking connection. Compatible with needleless IV sites designed for Luer-lock access, and Luer locking Foley Catheter sampling ports.