Gram's iodine solution stabilized

Supplier: HARDY DIAGNOSTICS CA

Synonyms: Lugol's iodine, Iodine and potassium iodide, Potassium triiodide

I008 I128
CA89407-620EA 37.1 CAD
CA89407-620 CA89407-622
Gram's iodine solution stabilized
Gram's iodine solution

A mordant used in the gram stain procedure. A stabilized, thicker iodine solution. Use with the intermediate decolorizer (Cat. no. D008) or fast decolorizer (Cat. no. D008F).


  • Ready to use
  • No ampules
  • A longer shelf life

Formula: I₃K
MW: 419.812 g/mol
MDL Number: MFCD00134128
CAS Number: 12298-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR